Rhodamine 123, 99+%, pure
Rhodamine 123, 99+%, pure
Rhodamine 123, 99+%, pure
Rhodamine 123, 99+%, pure
Thermo Scientific Chemicals
Rhodamine 123, 99+%, pure
CAS: 62669-70-9 | C21H19ClN2O4 | 398.843 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number FSA419091000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialRhodamine 123
Name Note99+%
CAS62669-70-9
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.