Pyridine, 99+%, extra pure, nonaqueous titration grade
Pyridine, 99+%, extra pure, nonaqueous titration grade
Pyridine, 99+%, extra pure, nonaqueous titration grade
Pyridine, 99+%, extra pure, nonaqueous titration grade
Thermo Scientific Chemicals
Pyridine, 99+%, extra pure, nonaqueous titration grade
CAS: 110-86-1 | C5H5N | 79.102 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
Packaging:
Glass Bottle
Catalog number FSA418530010
Price (MYR)
696.00
BOT
Add to cart
Quantity:
1 kg
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
696.00
BOT
Add to cart
Specifications
Chemical Name or MaterialPyridine
Melting Point-42.0°C
CAS110-86-1
Health Hazard 1Danger
Health Hazard 2GHS P Statement
Harmful in contact with skin.
Harmful if inhaled.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.