Pulegone, 92%, pract.
Pulegone, 92%, pract.
Pulegone, 92%, pract.
Pulegone, 92%, pract.
Thermo Scientific Chemicals
Pulegone, 92%, pract.
Pulegone, CAS # 89-82-7, is a compound found in the oils of certain plant species including peppermint, catnip, and pennyroyal. | CAS: 89-82-7 | C10H16O | 152.24 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA418501000100 gGlass bottle
Catalog number FSA418501000
Price (MYR)
795.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
795.00
EA
Add to cart
Specifications
Chemical Name or MaterialPulegone
CAS89-82-7
ColorColorless to Yellow
Density0.9310g/mL
EINECS Number201-943-2
View more
General description
• Pulegone is a p-menthane monoterpenoid and a known carcinogen
• Pulegone has a minty odor and taste at certain concentrations
Applications
• Pulegone may be useful as a natural insecticide
• Suitable for cell culture and in vivo research on toxicity, cellular redox state, and gene mutation
RUO – Research Use Only
General References:
  1. Zhang, Z.; Xie, Y.; Wang ,Y.; Lin, Z.; Wang, L.; Li, G. Toxicities of monoterpenes against housefly, Musca domestica L. (Diptera: Muscidae). Environ Sci Pollut Res Int. 2017, 31, 24708-24713.
  2. Schomburg, D.; Schomburg, I. 2013, (+)-pulegone reductase 1.3.1.81. In: Schomburg, D.; Schomburg, I. (eds) Class 1 Oxidoreductases. Springer Handbook of Enzymes. Springer, Berlin, Heidelberg.