Potassium metabisulfite, 97%, extra pure
Potassium metabisulfite, 97%, extra pure
Potassium metabisulfite, 97%, extra pure
Potassium metabisulfite, 97%, extra pure
Thermo Scientific Chemicals
Potassium metabisulfite, 97%, extra pure
CAS: 16731-55-8 | K2O5S2 | 222.31 g/mol
Have Questions?
Catalog NumberQuantityPackaging
FSA41827025025 gGlass bottle
Catalog number FSA418270250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium metabisulfite
Melting Point150.0°C
Name Note97%
CAS16731-55-8
ColorWhite to Yellow
View more
Reduction reagent
RUO – Research Use Only