Potassium iodate, 99.4+%, ACS reagent
Potassium iodate, 99.4+%, ACS reagent
Potassium iodate, 99.4+%, ACS reagent
Potassium iodate, 99.4+%, ACS reagent
Thermo Scientific Chemicals
Potassium iodate, 99.4+%, ACS reagent
CAS: 7758-05-6 | IKO3 | 214.00 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Plastic bottle
Catalog number FSA418241000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium iodate
CAS7758-05-6
ColorWhite
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
May cause respiratory irritation.
Causes serious eye irritation.
May intensify fire; oxidizer.
View more
Oxidation reagent
RUO – Research Use Only