Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
Thermo Scientific Chemicals
Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
CAS: 40949-94-8 | C6H18KNSi2 | 199.485 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number FSA418238000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS40949-94-8, 108-88-3
Specifications
Chemical Name or MaterialPotassium bis(trimethylsilyl)amide
ColorYellow
Name Note0.5M solution in toluene
CAS40949-94-8
Health Hazard 1Danger