View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialAcetic acid, potassium salt
pH6.5 to 9.0 (5% soln. at 25°C)
CAS127-08-2
Melting Point292.0°C