Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
Thermo Scientific Chemicals
Polyvinylsulfuric acid potassium salt, approx. M.W. 175,000
CAS: 26837-42-3 | (C2H3KO4S)n | NaN g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
Glass bottle
Catalog number FSA418141000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyvinylsulfuric acid potassium salt
ColorBeige-Brown to White
CAS26837-42-3
Physical FormCrystalline Powder
Molecular Formula(C2H3KO4S)n