Potassium hydrogen phthalate, 99+%
Potassium hydrogen phthalate, 99+%
Potassium hydrogen phthalate, 99+%
Potassium hydrogen phthalate, 99+%
Thermo Scientific Chemicals
Potassium hydrogen phthalate, 99+%
CAS: 877-24-7 | C8H5KO4 | 204.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA417955000
Price (MYR)
475.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
475.00
EA
Add to cart
Specifications
Chemical Name or MaterialPotassium hydrogen phthalate
Name Note99+%
CAS877-24-7
Melting Point295.0°C to 300.0°C
ColorRed