Phosphomolybdic Acid hydrate, ACS Reagent
Phosphomolybdic Acid hydrate, ACS Reagent
Phosphomolybdic Acid hydrate, ACS Reagent
Phosphomolybdic Acid hydrate, ACS Reagent
Thermo Scientific Chemicals
Phosphomolybdic Acid hydrate, ACS Reagent
CAS: 51429-74-4 | H3Mo12O40P | 1825.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
View all
Glass Bottle
Catalog number FSA417895000
Price (MYR)
3,064.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
3,064.00
EA
Add to cart
Specifications
Chemical Name or MaterialPhosphomolybdic Acid hydrate
ColorYellow
CAS51429-74-4
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Causes severe skin burns and eye damage.
May intensify fire; oxidizer.
View more
Oxidation reagent
RUO – Research Use Only