3-Phenylphenol, 90%
3-Phenylphenol, 90%
3-Phenylphenol, 90%
Thermo Scientific Chemicals
3-Phenylphenol, 90%
CAS: 580-51-8 | C12H10O | 170.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA417660250
Price (MYR)
1,998.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,998.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Phenylphenol
Name Note90%
CAS580-51-8
CAS Min %88.0
Health Hazard 1GHS Signal Word: Warning