p-Phenylenediamine dihydrochloride, 99+%
p-Phenylenediamine dihydrochloride, 99+%
p-Phenylenediamine dihydrochloride, 99+%
p-Phenylenediamine dihydrochloride, 99+%
Thermo Scientific Chemicals
p-Phenylenediamine dihydrochloride, 99+%
CAS: 624-18-0 | C6H8N2·2HCl | 181.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA417481000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialp-Phenylenediamine dihydrochloride
Name Note99+%
CAS624-18-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Toxic if inhaled.
Toxic in contact with skin.
Very toxic to aquatic life with long lasting effects.