Phenolphthalein, ACS reagent
Phenolphthalein, ACS reagent
Phenolphthalein, ACS reagent
Thermo Scientific Chemicals
Phenolphthalein, ACS reagent
CAS: 77-09-8 | C20H14O4 | 318.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA417185000
Price (MYR)
741.00
Bottle
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
741.00
Bottle
Specifications
Chemical Name or MaterialPhenolphthalein
Melting Point258.0°C to 263.0°C
CAS77-09-8
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
May cause cancer.
Suspected of causing genetic defects.
Suspected of damaging fertility.
View more
Indicator
RUO – Research Use Only