1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%
1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%
1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%
1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%
Thermo Scientific Chemicals
1,1,4,7,7-Pentamethyldiethylenetriamine, 98+%
CAS: 3030-47-5 | C9H23N3 | 173.3 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA416951000
Price (MYR)
450.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
450.00
BOT
Add to cart
Specifications
Chemical Name or Material1,1,4,7,7-Pentamethyldiethylenetriamine
CAS3030-47-5
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0