Peanut oil
Thermo Scientific Chemicals
Peanut oil
CAS: 8002-03-7
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA416855000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialPeanut oil
CAS8002-03-7
ColorYellow
Physical FormViscous Liquid
RTECS NumberRX2830000