Paraformaldehyde, 96%, extra pure
Paraformaldehyde, 96%, extra pure
Paraformaldehyde, 96%, extra pure
Paraformaldehyde, 96%, extra pure
Thermo Scientific Chemicals
Paraformaldehyde, 96%, extra pure
Paraformaldehyde, 96%, CH2O, CAS Number-30525-89-4, paraformaldehyde, paraform, oxymethylene, methylene oxide, methyl aldehyde, methanal, formol, formalin, formic aldehyde, oxomethane, 500g, CHEBI:16842, White, 260 deg.C, 1.03 (Air - 1.0), 71 deg.C, Extra Pure, Warning | CAS: 30525-89-4 | CH2O | 30.026 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
1 kg
Packaging:
View all
Plastic Bottle
Catalog number FSA416785000
Price (MYR)
375.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic Bottle
Request bulk or custom format
Price (MYR)
375.00
BOT
Add to cart
Specifications
Chemical Name or MaterialParaformaldehyde
Name NoteExtra Pure
CAS30525-89-4
Health Hazard 1Warning
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Suspected of causing cancer.
May cause an allergic skin reaction.
Harmful if inhaled.
Flammable in liquid state, upper limit 73%, lower limit 7.0%.