Paraffin, pure, wax, granular
Paraffin, pure, wax, granular
Paraffin, pure, wax, granular
Paraffin, pure, wax, granular
Thermo Scientific Chemicals
Paraffin, pure, wax, granular
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure | CAS: 8002-74-2 | CnH2n+2 | 341.451 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 kg
2 kg
Catalog number FSA416770100
Price (MYR)
2,397.00
EA
Add to cart
Quantity:
10 kg
Request bulk or custom format
Price (MYR)
2,397.00
EA
Add to cart
Specifications
Chemical Name or MaterialParaffin wax
CAS8002-74-2
Melting Point58.0°C to 62.0°C
ColorWhite
RTECS NumberRV0350000
View more
Paraffin wax, CnH2n+2, CAS Number-8002-74-2, propafenona inn-spanish, propafenona, propafenone-hcl, propafenonum inn-latin, propafenonum, propafenone inn:ban, propafenone inn, rythmol, propafenone, gnf-pf-4594, 2kg, 322.0 deg.C, CHEBI:63619, White, 232-315-6, 198 deg.C, Pure
RUO – Research Use Only