2-Hydroxy-5-nitrobenzaldehyde, 98+%
2-Hydroxy-5-nitrobenzaldehyde, 98+%
2-Hydroxy-5-nitrobenzaldehyde, 98+%
2-Hydroxy-5-nitrobenzaldehyde, 98+%
Thermo Scientific Chemicals
2-Hydroxy-5-nitrobenzaldehyde, 98+%
CAS: 97-51-8 | C7H5NO4 | 167.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA416180250
Price (MYR)
785.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
785.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Hydroxy-5-nitrobenzaldehyde
Name Note99%
CAS97-51-8
CAS Min %98.0
Health Hazard 1GHS Signal Word: Warning