4'-Nitroacetanilide, 99%
4'-Nitroacetanilide, 99%
4'-Nitroacetanilide, 99%
4'-Nitroacetanilide, 99%
Thermo Scientific Chemicals
4'-Nitroacetanilide, 99%
CAS: 104-04-1 | C8H8N2O3 | 180.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA415751000
Price (MYR)
1,990.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,990.00
BOT
Add to cart
Specifications
Chemical Name or Material4′-Nitroacetanilide
CAS104-04-1
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0