Nigrosine, pure, water soluble
Nigrosine, pure, water soluble
Nigrosine, pure, water soluble
Nigrosine, pure, water soluble
Thermo Scientific Chemicals
Nigrosine, pure, water soluble
CAS: 8005-03-6 | C22H14N6Na2O9S2 | 616.49 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA415685000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialNigrosine
Name Notepract.
CAS8005-03-6
TSCATSCA
Molecular FormulaC22H14N6Na2O9S2