Nickel phthalocyanine, 95%
Nickel phthalocyanine, 95%
Nickel phthalocyanine, 95%
Nickel phthalocyanine, 95%
Thermo Scientific Chemicals
Nickel phthalocyanine, 95%
CAS: 14055-02-8 | C32H16N8Ni | 571.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA415600250
Price (MYR)
1,188.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,188.00
BOT
Add to cart
Specifications
Chemical Name or MaterialNickel phthalocyanine
Name Note95%
CAS14055-02-8
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger