Nickel(II) acetate tetrahydrate, 99%, extra pure
Nickel(II) acetate tetrahydrate, 99%, extra pure
Nickel(II) acetate tetrahydrate, 99%, extra pure
Nickel(II) acetate tetrahydrate, 99%, extra pure
Thermo Scientific Chemicals
Nickel(II) acetate tetrahydrate, 99%, extra pure
CAS: 6018-89-9 | C4H14NiO8 | 248.84 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA415545000
Price (MYR)
565.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
565.00
BOT
Add to cart
Specifications
Chemical Name or MaterialNickel(II) acetate tetrahydrate
Name NoteExtra Pure
CAS6018-89-9
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger