alpha-Naphtholbenzein
alpha-Naphtholbenzein
alpha-Naphtholbenzein
alpha-Naphtholbenzein
Thermo Scientific Chemicals
alpha-Naphtholbenzein
Indicator | CAS: 145-50-6 | C27H18O2 | 374.44 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Catalog number FSA415321000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Materialα-Naphtholbenzein
CAS145-50-6
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and kee