Naphthol AS-BI phosphate, 93%
Naphthol AS-BI phosphate, 93%
Naphthol AS-BI phosphate, 93%
Naphthol AS-BI phosphate, 93%
Thermo Scientific Chemicals
Naphthol AS-BI phosphate, 93%
CAS: 1919-91-1 | C18H15BrNO6P | 452.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
Packaging:
Glass bottle
Catalog number FSA415311000
Price (MYR)
420.00
EA
Add to cart
Quantity:
100 mg
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
420.00
EA
Add to cart
Specifications
Chemical Name or MaterialNaphthol AS-BI phosphate
CAS1919-91-1
CAS Min %91.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0