Morpholine, 99+%, ACS reagent
Morpholine, 99+%, ACS reagent
Morpholine, 99+%, ACS reagent
Morpholine, 99+%, ACS reagent
Thermo Scientific Chemicals
Morpholine, 99+%, ACS reagent
CAS: 110-91-8 | C4H9NO | 87.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
Packaging:
View all
Glass Bottle
Catalog number FSA415165000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMorpholine
Name Note99+%
CAS110-91-8
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger