Mineral oil, pure
Thermo Scientific Chemicals
Mineral oil, pure
CAS: 8042-47-5 | C16H10N2Na2O7S2 | 452.363 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
6 mL
1 L
2.5 L
Catalog number FSA415080060
Price (MYR)
273.00
BOT
Add to cart
Quantity:
6 mL
Request bulk or custom format
Price (MYR)
273.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMineral oil
CAS8042-47-5
ColorColorless
Density0.8770g/mL
EINECS Number232-455-8