Dimethyl trisulfide, 98+%
Dimethyl trisulfide, 98+%
Dimethyl trisulfide, 98+%
Dimethyl trisulfide, 98+%
Thermo Scientific Chemicals
Dimethyl trisulfide, 98+%
CAS: 3658-80-8 | C2H6S3 | 126.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA415030050
Price (MYR)
506.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
506.00
BOT
Add to cart
Specifications
Chemical Name or MaterialDimethyl trisulfide
Name Note98+%
CAS3658-80-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Flammable liquid and vapor.