Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals
Thermo Scientific Chemicals
Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
2.5 L
Packaging:
View all
EcoSafPak™ Plastic Bottle
Catalog number FSA414885000
Price (MYR)
649.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
EcoSafPak™ Plastic Bottle
Request bulk or custom format
Price (MYR)
649.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl sulfoxide
Melting Point18.4°C
CAS67-68-5
CAS Min %99.9