Methyl DL-lactate, 99%
Methyl DL-lactate, 99%
Methyl DL-lactate, 99%
Methyl DL-lactate, 99%
Thermo Scientific Chemicals
Methyl DL-lactate, 99%
CAS: 547-64-8 | C4H8O3 | 104.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number FSA414480250
Price (MYR)
1,046.00
BOT
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (MYR)
1,046.00
BOT
Add to cart
Specifications
Chemical Name or MaterialMethyl DL-lactate
Name Note99%
CAS547-64-8
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Flammable liquid and vapor.