Methyl 4-iodobenzoate, 98%
Methyl 4-iodobenzoate, 98%
Methyl 4-iodobenzoate, 98%
Methyl 4-iodobenzoate, 98%
Thermo Scientific Chemicals
Methyl 4-iodobenzoate, 98%
CAS: 619-44-3 | C8H7IO2 | 262.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number FSA414460250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethyl 4-iodobenzoate
Name Note98%
CAS619-44-3
Health Hazard 1Warning
Health Hazard 2GHS H Statement
Toxic to aquatic life with long lasting effects.