Methylene di-p-phenyl diisocyanate, 98%, flakes
Methylene di-p-phenyl diisocyanate, 98%, flakes
Methylene di-p-phenyl diisocyanate, 98%, flakes
Methylene di-p-phenyl diisocyanate, 98%, flakes
Thermo Scientific Chemicals
Methylene di-p-phenyl diisocyanate, 98%, flakes
CAS: 101-68-8 | C15H10N2O2 | 250.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA414285000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialMethylene di-p-phenyl diisocyanate
Name Note99.5%
CAS101-68-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Suspected of causing cancer.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Harmf