3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%
3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%
3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%
3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%
Thermo Scientific Chemicals
3-Methyl-2-benzothiazolinone Hydrazone Hydrochloride Monohydrate, 98%
CAS: 38894-11-0 | HCl·H2O | 233.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
10 g
50 g
Packaging:
Glass bottle
Catalog number FSA414022500
Price (MYR)
10,221.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
10,221.00
EA
Add to cart
Specifications
Chemical Name or Material3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate
CAS38894-11-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0