2-(1-Methoxy)propyl acetate, 99%
2-(1-Methoxy)propyl acetate, 99%
2-(1-Methoxy)propyl acetate, 99%
2-(1-Methoxy)propyl acetate, 99%
Thermo Scientific Chemicals
2-(1-Methoxy)propyl acetate, 99%
CAS: 108-65-6 | C6H12O3 | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 kg
1 kg
Packaging:
View all
Plastic drum
Catalog number FSA413900050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 kg
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or Material2-(1-Methoxy)propyl acetate
CAS108-65-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0