Lithium stearate, 95%
Lithium stearate, 95%
Lithium stearate, 95%
Lithium stearate, 95%
Thermo Scientific Chemicals
Lithium stearate, 95%
CAS: 4485-12-5 | C18H35LiO2 | 290.41 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA413341000
Price (MYR)
771.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
771.00
BOT
Add to cart
Specifications
Chemical Name or MaterialLithium stearate
Name Note95%
CAS4485-12-5
Melting Point220°C