Lithium dodecyl sulfate, 99+%
Lithium dodecyl sulfate, 99+%
Lithium dodecyl sulfate, 99+%
Lithium dodecyl sulfate, 99+%
Thermo Scientific Chemicals
Lithium dodecyl sulfate, 99+%
CAS: 2044-56-6 | C12H25LiO4S | 272.33 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA413300250
Price (MYR)
1,257.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,257.00
BOT
Add to cart
Specifications
Absorbance(5% aq. soln.) (1 cm cell), (5% aq. soln.) (1 cm cell) at 0nm, 0.1 max. at 260nm, 0.1 max. at 280nm
Chemical Name or MaterialLithium dodecyl sulfate
ColorWhite
CAS2044-56-6
Health Hazard 1GHS Signal Word: Warning