Linoleic acid, 70%
Linoleic acid, 70%
Linoleic acid, 70%
Thermo Scientific Chemicals
Linoleic acid, 70%
CAS: 60-33-3 | C18H32O2 | 280.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number FSA413195000
Price (MYR)
655.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
655.00
BOT
Add to cart
Specifications
Chemical Name or MaterialLinoleic acid
Name Notetech., 60%
Acidity197 to 203mg KOH/g
CAS60-33-3
Health Hazard 1Warning