Isopropyl myristate, 96%
Isopropyl myristate, 96%
Isopropyl myristate, 96%
Isopropyl myristate, 96%
Thermo Scientific Chemicals
Isopropyl myristate, 96%
CAS: 110-27-0 | C17H34O2 | 270.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
1 kg
Packaging:
Glass bottle
Catalog number FSA412851000
Price (MYR)
249.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
249.00
BOT
Add to cart
Specifications
Chemical Name or MaterialIsopropyl myristate
Name Note96%
Acidity0.5mg KOH/g max.
CAS110-27-0
Health Hazard 2GHS H Statement: