Isopropyl methanesulfonate, 99%
Isopropyl methanesulfonate, 99%
Isopropyl methanesulfonate, 99%
Isopropyl methanesulfonate, 99%
Thermo Scientific Chemicals
Isopropyl methanesulfonate, 99%
CAS: 926-06-7 | C4H10O3S | 138.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
5 g
25 g
Packaging:
View all
Glass Bottle
Catalog number FSA412841000
Price (MYR)
5,266.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Price (MYR)
5,266.00
EA
Add to cart
Specifications
Chemical Name or MaterialIsopropyl methanesulfonate
Name Note99%
CAS926-06-7
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Harmful if swallowed.
Causes severe skin burns and eye damage.
Suspected of causing genetic defects.
Suspected of causing cancer.