Isopropanol, 99.5%, for spectroscopy ACS
Isopropanol, 99.5%, for spectroscopy ACS
Isopropanol, 99.5%, for spectroscopy ACS
Isopropanol, 99.5%, for spectroscopy ACS
Isopropanol, 99.5%, for spectroscopy ACS
CAS: 67-63-0 | C3H8O | 60.096 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
Packaging:
Glass Bottle
Catalog number FSA412790010
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropanol
Melting Point-89.5°C
Absorbance0.01 max. at 330nm, 0.01 max. at 400nm, 0.02 max. at 300nm, 0.03 max. at 275nm, 0.04 max. at 260nm, 0.08 max. at 245nm, 0.2 max. at 230nm, 0.4 max. at 220nm, 1 max. at 210nm
CAS67-63-0
Health Hazard 1Danger
View more
For UV/VIS spectroscopy
RUO – Research Use Only