2-Iodophenol, 98%
2-Iodophenol, 98%
2-Iodophenol, 98%
Thermo Scientific Chemicals
2-Iodophenol, 98%
CAS: 533-58-4 | C6H5IO | 220.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA412480250
Price (MYR)
789.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
789.00
BOT
Add to cart
Specifications
Chemical Name or Material2-Iodophenol
Name Note98%
CAS533-58-4
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning