4-Iodobenzonitrile, 99%, Thermo Scientific Chemicals
4-Iodobenzonitrile, 99%, Thermo Scientific Chemicals
4-Iodobenzonitrile, 99%, Thermo Scientific Chemicals
4-Iodobenzonitrile, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals
4-Iodobenzonitrile, 99%, Thermo Scientific Chemicals
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Catalog number FSA412380050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Iodobenzonitrile
Name Note99%
CAS3058-39-7
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning