2-Imidazolidone, 96%, anhydrous
2-Imidazolidone, 96%, anhydrous
2-Imidazolidone, 96%, anhydrous
2-Imidazolidone, 96%, anhydrous
Thermo Scientific Chemicals
2-Imidazolidone, 96%, anhydrous
CAS: 120-93-4 | C3H6N2O | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA412272500
Price (MYR)
639.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
639.00
EA
Add to cart
Specifications
Chemical Name or Material2-Imidazolidone
CAS120-93-4
CAS Min %95.0
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0