3'-Hydroxyacetophenone, 99+%
3'-Hydroxyacetophenone, 99+%
3'-Hydroxyacetophenone, 99+%
3'-Hydroxyacetophenone, 99+%
Thermo Scientific Chemicals
3'-Hydroxyacetophenone, 99+%
CAS: 121-71-1 | C8H8O2 | 136.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA411911000
Price (MYR)
2,577.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,577.00
BOT
Add to cart
Specifications
Chemical Name or Material3'-Hydroxyacetophenone
Name Note99+%
CAS121-71-1
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning