4-Hydrazinobenzoic acid, 98%
4-Hydrazinobenzoic acid, 98%
4-Hydrazinobenzoic acid, 98%
4-Hydrazinobenzoic acid, 98%
Thermo Scientific Chemicals
4-Hydrazinobenzoic acid, 98%
CAS: 619-67-0 | C7H8N2O2 | 152.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA411790050
Price (MYR)
276.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
276.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Hydrazinobenzoic acid
CAS619-67-0
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0