1,6-Hexanediamine, 60 wt % aqueous soln.
1,6-Hexanediamine, 60 wt % aqueous soln.
1,6-Hexanediamine, 60 wt % aqueous soln.
1,6-Hexanediamine, 60 wt % aqueous soln.
Thermo Scientific Chemicals
1,6-Hexanediamine, 60 wt % aqueous soln.
CAS: 124-09-4 | C6H16N2 | 116.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Plastic bottle
Catalog number FSA411562500
Price (MYR)
200.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
200.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 6-Hexanediamine
Name Note60 wt % aqueous soln.
CAS124-09-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement