Hexadecyltrimethylammonium hydroxide, 25% in methanol
Hexadecyltrimethylammonium hydroxide, 25% in methanol
Hexadecyltrimethylammonium hydroxide, 25% in methanol
Hexadecyltrimethylammonium hydroxide, 25% in methanol
Thermo Scientific Chemicals
Hexadecyltrimethylammonium hydroxide, 25% in methanol
CAS: 505-86-2 | C19H43NO | 301.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
Packaging:
Glass bottle
Catalog number FSA411420500
Price (MYR)
933.00
EA
Add to cart
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
933.00
EA
Add to cart
Specifications
Chemical Name or MaterialHexadecyltrimethylammonium hydroxide
ColorColorless to Yellow
Name Note25%in Methanol
CAS505-86-2
Health Hazard 1GHS Signal Word: Danger
View more
Phase transfer catalyst
RUO – Research Use Only