n-Heptane, 99+%, for spectroscopy
n-Heptane, 99+%, for spectroscopy
n-Heptane, 99+%, for spectroscopy
n-Heptane, 99+%, for spectroscopy
Thermo Scientific Chemicals
n-Heptane, 99+%, for spectroscopy
CAS: 142-82-5 | C7H16 | 100.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
Packaging:
View all
EcoSafPak™ Glass Bottle
Catalog number FSA411255000
Price (MYR)
446.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
EcoSafPak™ Glass Bottle
Request bulk or custom format
Price (MYR)
446.00
BOT
Add to cart
Specifications
Chemical Name or Materialn-Heptane
Melting Point-91.0°C
Absorbance0.01 max. at 240nm, 0.01 max. at 250nm, 0.01 max. at 260nm, 0.01 max. at 400nm, 0.05 max. at 230nm, 0.15 max. at 220nm, 0.4 max. at 210nm, 1 max. at 197nm, 1 max. at 200nm
CAS142-82-5
CAS Min %99.0
View more
For UV/VIS spectroscopy
RUO – Research Use Only