Guanosine, 99%
Guanosine, 99%
Guanosine, 99%
Thermo Scientific Chemicals
Guanosine, 99%
CAS: 118-00-3 | C10H13N5O5 | 283.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA411131000
Price (MYR)
852.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
852.00
EA
Add to cart
Specifications
Chemical Name or MaterialGuanosine
CAS118-00-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Toxic if swallowed.
Health Hazard 3GHS P Statement
Wash face,hands and any exposed skin thoroughly after handling.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.