Glycolic acid, 70%
Glycolic acid, 70%
Glycolic acid, 70%
Thermo Scientific Chemicals
Glycolic acid, 70%
CAS: 79-14-1 | C2H4O3 | 76.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA411032500
Price (MYR)
192.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
192.00
BOT
Add to cart
Specifications
Chemical Name or MaterialGlycolic acid
Name Note70% in water
CAS79-14-1
Melting Point10.0°C
Boiling Point113.0°C