Glycine hydrochloride, 98%
Glycine hydrochloride, 98%
Glycine hydrochloride, 98%
Glycine hydrochloride, 98%
Thermo Scientific Chemicals
Glycine hydrochloride, 98%
CAS: 6000-43-7 | C2H6ClNO2 | 111.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass Bottle
Catalog number FSA411011000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycine hydrochloride
Melting Point178.0°C to 182.0°C
Name Note99%
CAS6000-43-7
Health Hazard 1MOISTURE SENSITIVE